Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-3-nitropyrazolo[1,5-a]pyrimidine 95%

Supplier: AMBEED, INC
Warning

A244661-1G A244661-250MG A244661-25G A244661-5G
77153-136EA 12 USD InStock
77153-136 77153-138 77153-140 77153-142
5-Chloro-3-nitropyrazolo[1,5-a]pyrimidine 95%
5-Chloro-3-nitropyrazolo[1,5-a]pyrimidine
Precaución: for Research Use Only


Formula: C₆H₃ClN₄O₂
MDL Number: MFCD22415211
CAS Number: 1363380-51-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty