Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-Methyl 4-aminochroman-6-carboxylate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A246550-100MG A246550-250MG
77153-202EA 185 USD InStock
77153-202 77153-204
(R)-Methyl 4-aminochroman-6-carboxylate hydrochloride 95%
(R)-Methyl 4-aminochroman-6-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₄ClNO₃
MW: 243.69 g/mol
MDL Number: MFCD22392691
CAS Number: 1246509-59-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty