Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Asp(Edans)-OH 95%

Supplier: AMBEED, INC
Warning

A247536-100MG A247536-1G A247536-250MG
77153-208EA 65 USD InStock
77153-208 77153-210 77153-212
Fmoc-Asp(Edans)-OH 95%
Fmoc-Asp(Edans)-OH
Precaución: for Research Use Only


Formula: C₃₁H₂₉N₃O₈S
MW: 603.646 g/mol
MDL Number: MFCD02094140
CAS Number: 182253-73-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty