Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acotiamide hydrochloride trihydrate 99%

Supplier: AMBEED, INC
Warning

A254230-1G A254230-250MG A254230-50MG A254230-5G A254230-25G
77153-292EA 15 USD InStock
77153-292 77153-294 77153-296 77153-298 77685-723
Acotiamide hydrochloride trihydrate 99%
Acotiamide hydrochloride trihydrate
Precaución: for Research Use Only


Formula: C₂₁H₃₇ClN₄O₈S
MDL Number: MFCD28101055
CAS Number: 773092-05-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty