Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl (2-Chlorobenzyl)phosphonate 98%

Supplier: AMBEED, INC
Warning

A259443-100MG A259443-1G A259443-250MG A259443-5G
77153-390EA 7 USD InStock
77153-390 77153-392 77153-394 77153-396
Diethyl (2-Chlorobenzyl)phosphonate 98%
Diethyl (2-Chlorobenzyl)phosphonate
Precaución: for Research Use Only


Formula: C₁₁H₁₆ClO₃P
MDL Number: MFCD22574983
CAS Number: 29074-98-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty