Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SGX-523 98+%

Supplier: AMBEED, INC
Warning

A263510-10MG A263510-1MG A263510-2MG A263510-50MG A263510-5MG A263510-100MG
77153-510EA 45 USD InStock
77153-510 77153-512 77153-514 77153-516 77153-518 77685-752
SGX-523 98+%
SGX-523
Precaución: for Research Use Only


Formula: C₁₈H₁₃N₇S
MDL Number: MFCD16660190
CAS Number: 1022150-57-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty