Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,3R,4R,5R,6S)-6-Methyltetrahydro-2H-pyran-2,3,4,5-tetraol hydrate 98%

Supplier: AMBEED, INC
Warning

A269967-100G A269967-25G A269967-25MG A269967-500G
77153-620EA 34 USD InStock
77153-620 77153-622 77153-624 77153-626
(2R,3R,4R,5R,6S)-6-Methyltetrahydro-2H-pyran-2,3,4,5-tetraol hydrate 98%
(2R,3R,4R,5R,6S)-6-Methyltetrahydro-2H-pyran-2,3,4,5-tetraol hydrate
Precaución: for Research Use Only


Formula: C₆H₁₄O₆
MDL Number: MFCD00071591
CAS Number: 6155-35-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty