Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

LRRK2-IN-1 98%

Supplier: AMBEED, INC
Warning

A281167-10MG A281167-1MG A281167-5MG
77153-830EA 91 USD InStock
77153-830 77153-832 77153-834
LRRK2-IN-1 98%
LRRK2-IN-1
Precaución: for Research Use Only


Formula: C₃₁H₃₈N₈O₃
MDL Number: MFCD22683805
CAS Number: 1234480-84-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty