Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl 3,5-dimethoxybenzylphosphonate 95%

Supplier: AMBEED, INC
Warning

A283132-100MG A283132-1G A283132-250MG A283132-5G
77153-866EA 32 USD InStock
77153-866 77153-868 77153-870 77153-872
Diethyl 3,5-dimethoxybenzylphosphonate 95%
Diethyl 3,5-dimethoxybenzylphosphonate
Precaución: for Research Use Only


Formula: C₁₃H₂₁O₅P
MW: 288.27 g/mol
MDL Number: MFCD18252337
CAS Number: 108957-75-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty