Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Kestose 98+%

Supplier: AMBEED, INC
Warning

A283488-100MG A283488-1G A283488-250MG A283488-5G
77153-874EA 67 USD InStock
77153-874 77153-876 77153-878 77153-880
1-Kestose 98+%
1-Kestose
Precaución: for Research Use Only


Formula: C₁₈H₃₂O₁₆
MDL Number: MFCD00142647
CAS Number: 470-69-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty