Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bicyclo[4.2.0]octa-1,3,5-triene-3-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A290256-100MG A290256-1G
77153-976EA 36 USD InStock
77153-976 77153-978
Bicyclo[4.2.0]octa-1,3,5-triene-3-carbaldehyde 97%
Bicyclo[4.2.0]octa-1,3,5-triene-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₉H₈O
MDL Number: MFCD09842341
CAS Number: 112892-88-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty