Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazole-3,5-dicarbonitrile 98%

Supplier: AMBEED, INC
Danger

A290645-100MG A290645-250MG
77153-984EA 45 USD InStock
77153-984 77153-986
1H-Pyrazole-3,5-dicarbonitrile 98%
1H-Pyrazole-3,5-dicarbonitrile
Precaución: for Research Use Only


Formula: C₅H₂N₄
MDL Number: MFCD28168072
CAS Number: 847573-68-6
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty