Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-fluorobenzenesulphonyl chloride 98%

Supplier: AMBEED, INC
Danger

A292472-1G A292472-250MG A292472-25G A292472-5G
77154-040EA 10 USD InStock
77154-040 77154-042 77154-044 77154-046
5-Chloro-2-fluorobenzenesulphonyl chloride 98%
5-Chloro-2-fluorobenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₆H₃Cl₂FO₂S
MW: 229.06
MDL Number: MFCD03094221
CAS Number: 351003-49-1
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty