Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nexturastat A 98%

Supplier: AMBEED, INC
Warning

A293543-1MG A293543-250MG A293543-50MG A293543-5MG
77154-070EA 15 USD InStock
77154-070 77154-072 77154-074 77154-076
Nexturastat A 98%
Nexturastat A
Precaución: for Research Use Only


Formula: C₁₉H₂₃N₃O₃
MDL Number: MFCD28099804
CAS Number: 1403783-31-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty