Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-Glu-OFm 95%

Supplier: AMBEED, INC
Warning

A302086-100MG A302086-250MG
77154-190EA 71 USD InStock
77154-190 77154-192
Fmoc-D-Glu-OFm 95%
Fmoc-D-Glu-OFm
Precaución: for Research Use Only


Formula: C₃₄H₂₉NO₆
MW: 547.60 g/mol
MDL Number: MFCD02094381
CAS Number: 252049-17-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty