Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

YM155 ≥98%

Supplier: AMBEED, INC
Warning

A307284-10MG A307284-1MG A307284-5MG A307284-50MG
77154-286EA 33 USD InStock
77154-286 77154-288 77154-290 77685-878
YM155 ≥98%
YM155
Precaución: for Research Use Only


Formula: C₂₀H₁₉BrN₄O₃
MDL Number: MFCD11983133
CAS Number: 781661-94-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty