Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Azaspiro[bicyclo[3.2.1]octane-3,2'-[1,3]dioxolane] 95%

Supplier: AMBEED, INC
Warning

A310336-100MG A310336-250MG
77154-346EA 105 USD InStock
77154-346 77154-348
8-Azaspiro[bicyclo[3.2.1]octane-3,2'-[1,3]dioxolane] 95%
8-Azaspiro[bicyclo[3.2.1]octane-3,2'-[1,3]dioxolane]
Precaución: for Research Use Only


Formula: C₉H₁₅NO₂
MDL Number: MFCD20661702
CAS Number: 13375-57-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty