Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-3-Fluorocinnamic acid 98%

Supplier: AMBEED, INC
Danger

A313110-25G A313110-5G
77154-420EA 24 USD InStock
77154-420 77154-422
trans-3-Fluorocinnamic acid 98%
trans-3-Fluorocinnamic acid
Precaución: for Research Use Only


Formula: C₉H₇FO₂
MW: 166.15
MDL Number: MFCD00004383
CAS Number: 20595-30-6
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty