Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(tert-Butoxycarbonyl)-DL-tryptophan 97%

Supplier: AMBEED, INC
Warning

A321815-100G A321815-10G A321815-1G A321815-25G A321815-500G A321815-5G
77154-524EA 51 USD InStock
77154-524 77154-526 77154-528 77154-530 77154-532 77154-534
N-(tert-Butoxycarbonyl)-DL-tryptophan 97%
N-(tert-Butoxycarbonyl)-DL-tryptophan
Precaución: for Research Use Only


Formula: C₁₆H₂₀N₂O₄
MDL Number: MFCD00190836
CAS Number: 112525-72-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty