Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SU-5416 98%

Supplier: AMBEED, INC
Warning

A325242-10MG A325242-1MG A325242-50MG A325242-5MG
77154-592EA 24 USD InStock
77154-592 77154-594 77154-596 77154-598
SU-5416 98%
SU-5416
Precaución: for Research Use Only


Formula: C₁₅H₁₄N₂O
MDL Number: MFCD01940922
CAS Number: 204005-46-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty