Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(triphenylphosphine)cobalt dichloride 97%

Supplier: AMBEED, INC
Warning

A329926-1G A329926-25G A329926-5G A329926-100G
77154-680EA 5 USD InStock
77154-680 77154-682 77154-684 77685-944
Bis(triphenylphosphine)cobalt dichloride 97%
Bis(triphenylphosphine)cobalt dichloride
Precaución: for Research Use Only


Formula: C₃₆H₃₀Cl₂CoP₂
MW: 654.41 g/mol
MDL Number: MFCD00010010
CAS Number: 14126-40-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty