Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Benzyloxycarbonyl-3-(Fmoc-amino)-L-alanine 98%

Supplier: AMBEED, INC
Warning

A333107-10G A333107-1G A333107-250MG A333107-5G
77154-736EA 50 USD InStock
77154-736 77154-738 77154-740 77154-742
N-Benzyloxycarbonyl-3-(Fmoc-amino)-L-alanine 98%
N-Benzyloxycarbonyl-3-(Fmoc-amino)-L-alanine
Precaución: for Research Use Only


Formula: C₂₆H₂₄N₂O₆
MW: 460.48 g/mol
MDL Number: MFCD00274016
CAS Number: 142855-80-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty