Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl alcohol 96%

Supplier: AMBEED, INC
Danger

A336659-100MG A336659-1G A336659-250MG
77154-808EA 50 USD InStock
77154-808 77154-810 77154-812
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl alcohol 96%
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl alcohol
Precaución: for Research Use Only


Formula: C₉H₇F₃N₂O
MDL Number: MFCD19442582
CAS Number: 87736-88-7
UN: 2924
ADR: 3,8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty