Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dimethyl-N-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propanamide 98+%

Supplier: AMBEED, INC
Warning

A344445-100MG A344445-1G A344445-250MG A344445-5G
77154-900EA 9 USD InStock
77154-900 77154-902 77154-904 77154-906
2,2-Dimethyl-N-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propanamide 98+%
2,2-Dimethyl-N-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propanamide
Precaución: for Research Use Only


Formula: C₁₇H₂₆BNO₃
MW: 303.20 g/mol
MDL Number: MFCD09842849
CAS Number: 1409999-54-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty