Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rebaudioside A 98%

Supplier: AMBEED, INC
Warning

A344662-100G A344662-100MG A344662-25G A344662-25MG A344662-500G A344662-5G
77154-910EA 95 USD InStock
77154-910 77154-912 77154-914 77154-916 77154-918 77154-920
Rebaudioside A 98%
Rebaudioside A
Precaución: for Research Use Only


Formula: C₄₄H₇₀O₂₃
MDL Number: MFCD02183463
CAS Number: 58543-16-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty