Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-dihydro-2-benzofuran-1-one 98%

Supplier: AMBEED, INC
Warning

A349597-100MG A349597-1G A349597-250MG A349597-5G
77154-966EA 22 USD InStock
77154-966 77154-968 77154-970 77154-972
5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-dihydro-2-benzofuran-1-one 98%
5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-dihydro-2-benzofuran-1-one
Precaución: for Research Use Only


Formula: C₁₄H₁₇BO₄
MDL Number: MFCD18251723
CAS Number: 862081-37-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty