Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-2-methylphenylacetic acid 95%

Supplier: AMBEED, INC
Warning

A352134-250MG A352134-100MG
77154-994EA 119 USD InStock
77154-994 77686-002
4-Methoxy-2-methylphenylacetic acid 95%
4-Methoxy-2-methylphenylacetic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
MW: 180.21 g/mol
MDL Number: MFCD02664778
CAS Number: 942-97-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty