Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

C-(5-Methyl[1,2,4]oxadiazol-3-yl)methylamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A360300-100MG A360300-1G A360300-250MG A360300-25G A360300-5G
77155-098EA 21 USD InStock
77155-098 77155-100 77155-102 77155-104 77155-106
C-(5-Methyl[1,2,4]oxadiazol-3-yl)methylamine hydrochloride 95%
C-(5-Methyl[1,2,4]oxadiazol-3-yl)methylamine hydrochloride
Precaución: for Research Use Only


Formula: C₄H₈ClN₃O
MW: 149.58 g/mol
MDL Number: MFCD11506367
CAS Number: 1184986-84-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty