Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

KY02111 98+%

Supplier: AMBEED, INC
Warning

A360800-10MG A360800-1MG A360800-50MG A360800-5MG
77155-112EA 16 USD InStock
77155-112 77155-114 77155-116 77155-118
KY02111 98+%
KY02111
Precaución: for Research Use Only


Formula: C₁₈H₁₇ClN₂O₃S
MW: 376.86 g/mol
MDL Number: MFCD08483062
CAS Number: 1118807-13-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty