Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-(-)-Leucine 98%

Supplier: AMBEED, INC
Warning

A362814-100G A362814-10G A362814-25G A362814-500G A362814-5G
77155-146EA 50 USD InStock
77155-146 77155-148 77155-150 77155-152 77155-154
D-(-)-Leucine 98%
D-(-)-Leucine
Precaución: for Research Use Only


Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 226
MDL Number: MFCD00063088
CAS Number: 328-38-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty