Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(3-Chlorophenyl)-1,3,4-thiadiazol-2-amine 95%

Supplier: AMBEED, INC
Warning

A363950-250MG A363950-25G
77155-158EA 10 USD InStock
77155-158 77155-160
5-(3-Chlorophenyl)-1,3,4-thiadiazol-2-amine 95%
5-(3-Chlorophenyl)-1,3,4-thiadiazol-2-amine
Precaución: for Research Use Only


Formula: C₈H₆ClN₃S
MDL Number: MFCD02664059
CAS Number: 70057-67-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty