Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',3'-Dideoxy-2'3'-didehydroadenosine 95%

Supplier: AMBEED, INC
Warning

A366641-1MG A366641-25MG
77155-188EA 5 USD InStock
77155-188 77155-190
2',3'-Dideoxy-2'3'-didehydroadenosine 95%
2',3'-Dideoxy-2'3'-didehydroadenosine
Precaución: for Research Use Only


Formula: C₁₀H₁₁N₅O₂
MDL Number: MFCD00040965
CAS Number: 7057-48-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty