Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethyl-3-methylimidazolium methanesulphonate 98%

Supplier: AMBEED, INC
Warning

A373767-100G A373767-10G A373767-1G A373767-25G A373767-5G
77155-228EA 104 USD InStock
77155-228 77155-230 77155-232 77155-234 77155-236
1-Ethyl-3-methylimidazolium methanesulphonate 98%
1-Ethyl-3-methylimidazolium methanesulphonate
Precaución: for Research Use Only


Formula: C₇H₁₄N₂O₃S
MW: 206.27 g/mol
MDL Number: MFCD06798171
CAS Number: 145022-45-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty