Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,5-Trifluorobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A381386-1G A381386-5G A381386-25G
77155-310EA 9 USD InStock
77155-310 77155-312 77686-064
2,4,5-Trifluorobenzonitrile 98%
2,4,5-Trifluorobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₂F₃N
MDL Number: MFCD00013289
CAS Number: 98349-22-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty