Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

LY-2874455 ≥98%

Supplier: AMBEED, INC
Warning

A382878-10MG A382878-1MG A382878-25MG A382878-5MG A382878-50MG
77155-338EA 53 USD InStock
77155-338 77155-340 77155-342 77155-344 77686-066
LY-2874455 ≥98%
LY-2874455
Precaución: for Research Use Only


Formula: C₂₁H₁₉Cl₂N₅O₂
MW: 444.32 g/mol
MDL Number: MFCD22124884
CAS Number: 1254473-64-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty