Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-((tert-butoxycarbonyl)amino)-2-(2-chlorophenyl)acetic acid 98%

Supplier: AMBEED, INC
Warning

A389853-250MG A389853-5G A389853-1G
77155-440EA 32 USD InStock
77155-440 77155-442 77686-081
(R)-2-((tert-butoxycarbonyl)amino)-2-(2-chlorophenyl)acetic acid 98%
(R)-2-((tert-butoxycarbonyl)amino)-2-(2-chlorophenyl)acetic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₆ClNO₄
MDL Number: MFCD03426364
CAS Number: 1212602-23-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty