Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ralimetinib dimesylate ≥98%

Supplier: AMBEED, INC
Warning

A413122-10MG A413122-1MG A413122-5MG
77155-740EA 34 USD InStock
77155-740 77155-742 77155-744
Ralimetinib dimesylate ≥98%
Ralimetinib dimesylate
Precaución: for Research Use Only


Formula: C₂₆H₃₇FN₆O₆S₂
MW: 612.74 g/mol
MDL Number: MFCD28101032
CAS Number: 862507-23-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty