Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

OSI-027 98%

Supplier: AMBEED, INC
Warning

A419082-10MG A419082-1MG A419082-50MG A419082-5MG A419082-100MG
77155-792EA 105 USD InStock
77155-792 77155-794 77155-796 77155-798 77686-157
OSI-027 98%
OSI-027
Precaución: for Research Use Only


Formula: C₂₁H₂₂N₆O₃
MDL Number: MFCD20275101
CAS Number: 936890-98-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty