Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Boc-amino)-3-fluoro-phenol 97%

Supplier: AMBEED, INC
Warning

A423103-10G A423103-1G A423103-5G
77155-826EA 304 USD InStock
77155-826 77155-828 77155-830
4-(Boc-amino)-3-fluoro-phenol 97%
4-(Boc-amino)-3-fluoro-phenol
Precaución: for Research Use Only


Formula: C₁₁H₁₄FNO₃
MW: 227.23 g/mol
MDL Number: MFCD19382482
CAS Number: 911297-02-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty