Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-(Diethylamino)-4-(hydroxymethyl)coumarin 95%

Supplier: AMBEED, INC
Warning

A426161-100MG A426161-1G A426161-50MG A426161-250MG
77155-868EA 158 USD InStock
77155-868 77155-870 77155-872 77613-320
7-(Diethylamino)-4-(hydroxymethyl)coumarin 95%
7-(Diethylamino)-4-(hydroxymethyl)coumarin
Precaución: for Research Use Only


Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
MDL Number: MFCD03424352
CAS Number: 54711-38-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty