Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-tert-Butylbenzenesulfonamide 96%

Supplier: AMBEED, INC
Warning

A429277-10G A429277-1G A429277-25G
77155-904EA 155 USD InStock
77155-904 77155-906 77155-908
N-tert-Butylbenzenesulfonamide 96%
N-tert-Butylbenzenesulphonamide
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO₂S
MDL Number: MFCD00462465
CAS Number: 2512-24-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty