Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloro-3-fluoro-4-iodobenzene 97%

Supplier: AMBEED, INC
Warning

A432653-100G A432653-25G A432653-500G A432653-5G
77155-934EA 47 USD InStock
77155-934 77155-936 77155-938 77155-940
1-Chloro-3-fluoro-4-iodobenzene 97%
1-Chloro-3-fluoro-4-iodobenzene
Precaución: for Research Use Only


Formula: C₆H₃ClFI
MDL Number: MFCD00079729
CAS Number: 6797-79-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty