Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3,5-difluorophenol 98%

Supplier: AMBEED, INC
Warning

A438641-100G A438641-1G A438641-25G A438641-5G
77156-008EA 519 USD InStock
77156-008 77156-010 77156-012 77156-014
4-Bromo-3,5-difluorophenol 98%
4-Bromo-3,5-difluorophenol
Precaución: for Research Use Only


Formula: C₆H₃BrF₂O
MW: 208.99
MDL Number: MFCD08062385
CAS Number: 130191-91-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty