Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(Trifluoromethylthio)phthalimide 98%

Supplier: AMBEED, INC
Warning

A442652-100MG A442652-1G A442652-5G
77156-076EA 14 USD InStock
77156-076 77156-078 77594-386
N-(Trifluoromethylthio)phthalimide 98%
N-(Trifluoromethylthio)phthalimide
Precaución: for Research Use Only


Formula: C₉H₄F₃NO₂S
MW: 247.19 g/mol
MDL Number: MFCD18449627
CAS Number: 719-98-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty