Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Decyl-3-methylimidazoliumtetrafluoroborate 98%

Supplier: AMBEED, INC
Warning

A447119-100G A447119-10G A447119-5G
77156-096EA 101 USD InStock
77156-096 77156-098 77156-100
1-Decyl-3-methylimidazoliumtetrafluoroborate 98%
1-Decyl-3-methylimidazoliumtetrafluoroborate
Precaución: for Research Use Only


Formula: C₁₄H₂₇BF₄N₂
MDL Number: MFCD09038853
CAS Number: 244193-56-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty