Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluorobenzenesulfonyl chloride 98%

Supplier: AMBEED, INC
Danger

A462045-10G A462045-25G A462045-5G
77156-206EA 13 USD InStock
77156-206 77156-208 77156-210
3,5-Difluorobenzenesulfonyl chloride 98%
3,5-Difluorobenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₆H₃ClF₂O₂S
MDL Number: MFCD02091378
CAS Number: 210532-25-5
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty