Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-methoxybenzaldehyde 98%

Supplier: AMBEED, INC
Danger

A469165-1G A469165-25G A469165-5G
77156-296EA 15 USD InStock
77156-296 77156-298 77156-300
2-Chloro-3-methoxybenzaldehyde 98%
2-Chloro-3-methoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₇ClO₂
MW: 170.5930 g/mol
MDL Number: MFCD07369787
CAS Number: 54881-49-1
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty