Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Naphthyl trifluoromethanesulfonate 98%

Supplier: AMBEED, INC
Danger

A470740-1G A470740-25G A470740-5G
77156-314EA 22 USD InStock
77156-314 77156-316 77156-318
2-Naphthyl trifluoromethanesulfonate 98%
2-Naphthyl trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₁₁H₇F₃O₃S
MDL Number: MFCD00192341
CAS Number: 3857-83-8
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty