Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methoxy-N-methyl-2-furancarboxamide 98%

Supplier: AMBEED, INC
Warning

A471049-1G A471049-250MG A471049-5G
77156-320EA 48 USD InStock
77156-320 77156-322 77156-324
N-Methoxy-N-methyl-2-furancarboxamide 98%
N-Methoxy-N-methyl-2-furancarboxamide
Precaución: for Research Use Only


Formula: C₇H₉NO₃
MDL Number: MFCD06804576
CAS Number: 95091-92-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty