Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trichlorobenzenesulfonyl chloride 98%

Supplier: AMBEED, INC
Danger

A479374-10G A479374-1G A479374-25G A479374-5G
77156-404EA 30 USD InStock
77156-404 77156-406 77156-408 77156-410 77156-412
2,4,6-Trichlorobenzenesulfonyl chloride 98%
2,4,6-Trichlorobenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₆H₂Cl₄O₂S
MDL Number: MFCD00052973
CAS Number: 51527-73-2
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty